Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Bis(2-propynyl) Ether 98.0+%, TCI America™
SDP

Catalog No. D45815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45815G 5 g
D458125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45815G Supplier TCI America Supplier No. D45815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 126422-57-9
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
InChI Key ZQNBDEOGKNZIQM-UHFFFAOYSA-N
Synonym 3-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]-1-propyne, 4,7,10-Trioxa-1,12-tridecadiyne
PubChem CID 11008566
IUPAC Name 3-[2-(2-prop-2-ynoxyethoxy)ethoxy]prop-1-yne
SMILES C#CCOCCOCCOCC#C

Specifications

Color Yellow
Boiling Point 103°C
Quantity 5 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Bis(2-propynyl) Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.