Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Butyl Methyl Ether 99.0+%, TCI America™
SDP

Catalog No. D467625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D467625ML 25 mL
D4676500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D467625ML Supplier TCI America Supplier No. D467625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7382-32-3
Molecular Formula C9H20O3
Molecular Weight (g/mol) 176.256
MDL Number MFCD00518661
InChI Key HYLLZXPMJRMUHH-UHFFFAOYSA-N
Synonym 1-[2-(2-Methoxyethoxy)ethoxy]butane
PubChem CID 81861
IUPAC Name 1-[2-(2-methoxyethoxy)ethoxy]butane
SMILES CCCCOCCOCCOC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 176.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Butyl Methyl Ether
EINECSNumber (7)-1321
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.