Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethylene glycol monoethyl ether acetate

Catalog No. 5375271L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
4 L
18 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-752-71L 1 L
53-752-74L 4 L
53-752-718L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 53-752-71L Supplier Sigma Aldrich Supplier No. 5375271L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethylene glycol monoethyl ether acetate

Specifications

CAS 112-15-2
Melting Point -25°C (lit.)
Density 1.011 g/mL (at 25°C (literature))
Boiling Point 218°C to 219°C (lit.)
Molecular Formula C8H16O4
Refractive Index n20/D 1.421 (literature)
Linear Formula C2H5OCH2CH2OCH2CH2CO2CH3
MDL Number MFCD00041928
Quantity 1 L
Synonym DE Acetate; 2-(2-Ethoxyethoxy)ethyl acetate; Ethyldiglycol acetate
Molecular Weight (g/mol) 176.21
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.