Learn More
Diethylene glycol monomethyl ether, 98%
Description
Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.
Solubility
Fully miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
Specifications
Specifications
| Melting Point | -70°C |
| Density | 1.01 |
| Quantity | 2500 mL |
| Flash Point | 83°C (181°F) |
| Odor | Light |
| Refractive Index | 1.426 |
| Linear Formula | CH3OCH2CH2OCH2CH2OH |
| Beilstein | 1697812 |
| Merck Index | 14,6037 |
| Boiling Point | 194°C |
| Show More |
Safety and Handling
| Hazard Category | H227-H311-H319-H361d |
| Hazard Statement | GHS H Statement H361-H227 Suspected of damaging fertility or the unborn child. Combustible liquid. |
| Precautionary Statement | P201-P202-P210-P233-P240-P243-P264b-P280-P302+P352-P305+P351+P338-P308+P313-P312-P362-P374-P501c |
| EINECSNumber | 203-906-6 |
| RTECSNumber | KL6125000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.