Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Monomethyl Ether Methacrylate (stabilized with MEHQ) 97.0+%, TCI America™
SDP

Catalog No. D153125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D153125ML 25 mL
D1531500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D153125ML Supplier TCI America Supplier No. D153125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 45103-58-0
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00059330
InChI Key DAVVKEZTUOGEAK-UHFFFAOYSA-N
Synonym Methacrylic Acid 2-(2-Methoxyethoxy)ethyl Ester, 2-(2-Methoxyethoxy)ethyl Methacrylate, Methyl Carbitol Methacrylate
PubChem CID 170752
IUPAC Name 2-(2-methoxyethoxy)ethyl 2-methylprop-2-enoate
SMILES COCCOCCOC(=O)C(C)=C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 188.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monomethyl Ether Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1049
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.