Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene Glycol Monophenyl Ether 98.0+%, TCI America™
SDP

Catalog No. D5139500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5139500G 500 g
D513925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5139500G Supplier TCI America Supplier No. D5139500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104-68-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00045989
InChI Key ZUAURMBNZUCEAF-UHFFFAOYSA-N
Synonym 2-(2-Phenoxyethoxy)ethanol, Phenyl Diglycol
PubChem CID 7715
IUPAC Name 2-(2-phenoxyethoxy)ethan-1-ol
SMILES OCCOCCOC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 298°C
Quantity 500 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monophenyl Ether
EINECSNumber (3)-0563
RTECSNumber KM0875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.