Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylenetriaminepentaacetic Acid, Pentasodium Salt, 41% Solution in Water, Spectrum™ Chemical
SDP

Catalog No. 18602202 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-202 500 mL Poly Bottle
18-602-201 100 mL Poly Bottle
18-602-203 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-602-202 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1156500ML
Add to Cart
Add to Cart

D1156, 140-01-2, C14H18N3Na5O10

Spectrum™ Chemical Diethylenetriaminepentaacetic Acid, Pentasodium Salt, 41 Percent Solution in Water is also known under the abbreviation DETA and is a colorless liquid that is hygroscopic and soluble in both water and organic solvent that are polarized. Used as a tridentate ligand forming complex in coordination chemistry, it is also used for the extraction of gases that are acid in the oil industry.

Chemical Identifiers

CAS 140-01-2
Molecular Formula C14H18N3Na5O10
Molecular Weight (g/mol) 503.26
MDL Number MFCD00051016
InChI Key LQPLDXQVILYOOL-UHFFFAOYSA-I
IUPAC Name pentasodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 mL
Percent Purity 39 to 41%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.