Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylenetriaminepentaacetic Dianhydride 98.0+%, TCI America™
SDP

Catalog No. D242425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D242425G 25 g
D24245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D242425G Supplier TCI America Supplier No. D242425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23911-26-4
Molecular Formula C14H19N3O8
Molecular Weight (g/mol) 357.319
MDL Number MFCD00010697
InChI Key RAZLJUXJEOEYAM-UHFFFAOYSA-N
Synonym dtpa-anhydride, cyclic dtpa anhydride, 2-bis 2-2,6-dioxomorpholino ethyl amino acetic acid, unii-wnm9rlh5x9, wnm9rlh5x9, diethylenetriaminepentaacetic dianhydride, diethylenetriaminepentaacetic acid dianhydride, cdtpaa, glycine, n,n-bis 2-2,6-dioxo-4-morpholinyl ethyl, bis 2-2,6-dioxomorpholin-4-yl ethyl amino acetic acid
PubChem CID 100825
IUPAC Name 2-[bis[2-(2,6-dioxomorpholin-4-yl)ethyl]amino]acetic acid
SMILES C1C(=O)OC(=O)CN1CCN(CCN2CC(=O)OC(=O)C2)CC(=O)O

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 25 g
Formula Weight 357.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylenetriaminepentaacetic Dianhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.