Learn More
Diethylstilbestrol, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 169°C to 175°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Quantity | 50 g |
| Beilstein | 06, III, 5619 |
| Merck Index | 15, 3148 |
| IUPAC Name | 4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol |
| Formula Weight | 268.34 |
| Percent Purity | ≥98.5% |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement: Suspected of causing cancer. May damage the unborn child. |
| Precautionary Statement | GHS P Statement: Use personal protective equipment as required. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 200-278-5 |
| RTECSNumber | WJ5600000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.