Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylthiocarbamoyl chloride, Thermo Scientific Chemicals

Catalog No. AC465920050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465920050 5 g
AC465920250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465920050 Supplier Thermo Scientific Chemicals Supplier No. 465920050
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-11-9
Molecular Formula C5H10ClNS
Molecular Weight (g/mol) 151.65
MDL Number MFCD00022106
InChI Key HUUSTUALCPTCGJ-UHFFFAOYSA-N
IUPAC Name (chloromethanethioyl)diethylamine
SMILES CCN(CC)C(Cl)=S

Specifications

Alpha Vector DIETHYLCARBAMOTHIOICCHLORIDE
Melting Point 45°C to 50°C
Color White to Yellow
Boiling Point 113°C
Flash Point 113°C
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
UN Number 3261
Sensitivity Moisture sensitive
Formula Weight 151.66
Physical Form Solid
Chemical Name or Material Diethylthiocarbamoyl chloride
Show More Show Less
Hazard Category GHS05: Corrosive
ShelfLife 2 years
storageNote1 normal conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.