Learn More
Enzo Life Sciences DIF-1 (100µg). CAS: 111050-72-7
SDP
Supplier:  Enzo Life Sciences BMLGR3240100
Encompass_Preferred
Supresses cell growth and promotes retinoic acid-induced cell differentiation in HL-60 cells. It is a known inhibitor of Wnt/β-Catenin signaling pathway. Alternative name: 1-(3,5-Dichloro-2,6-dihydroxy-4-methoxyphenyl) hexan-1-one, Differentiation-inducing factor-1. Purity: ≥98% (HPLC). Solubility: Soluble in ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0976
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.