Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Difco™ Brain Heart Infusion, without dextrose

Catalog No. DF0502085 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0502-08-5 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0502-08-5 Supplier Gibco™ Supplier No. 250220

missing translation for 'validMainframeMsg'

A general-purpose medium used in the cultivation of fastidous and nonfastidious microorganisms, including aerobic and anaerobic bacteria

Difco Brain Heart Infusion, without dextrose, is an animal origin (AO) enzymatic digest of bovine and porcine animal proteins. Difco Brain Heart Infusion, without dextrose, is a microbiological culture medium used for cultivating fastidious microorganisms, including streptococci, pneumococci, and meningococci.

Features:
• Widely used in various microbiological studies

Applications of Difco Brain Heart Infusion, without dextrose, include:
• For food testing and the verification of fecal streptococci
• As a medium for use in the preparation of microdilution trays for antimicrobial susceptibility testing by the broth microdilution procedure
• Recommended for the growth of most ATCC strains of Pasteurella multocida; it has been cited in many studies on P. multocida fowl cholera vaccines

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Form Dehydrated
Product Line Difco
Quantity 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.