Learn More
BD Difco™ Chocolate Agar Base (GC Medium)
Catalog No. DF0289075 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
2 kg
Catalog No. Quantity
DF0289-07-5 2 kg
1 options
This item is not returnable. View return policy
Catalog No. DF0289-07-5 Supplier BD Supplier No. 228920
This item is not returnable. View return policy
For use in cultivation of N. gonorrhoeae and other fastidious organisms
TRUSTED_SUSTAINABILITY
Specifications
Form Dehydrated
Quantity 2 kg