Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Fluid Lactose Medium

Catalog No. DF0004088 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10,000 g
2000 g
500 g
Packaging Type:
Bottle
Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0004-08-8 10,000 g Pail
DF0004-17-7 500 g Bottle
DF0004-07-9 2000 g Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. DF0004-08-8 Supplier BD Supplier No. 242000
Add to Cart
Add to Cart

For cultivation of Salmonella and coliform organisms in water, foods, dairy and pharmaceutical products

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, SMD
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Lactose Broth
Product Type Dehydrated Culture Media
Formulation Beef extract 3g/L, Peptone 5g/L, Lactose 5g/L
For Use With (Application) Cultivation of Salmonella and Coliform organisms in Water, foods, dairy and pharmaceutical products
Packaging Type Pail
Quantity 10,000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.