Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium

Catalog No. DF0256083 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10,000 g
2000 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0256-08-3 10,000 g
DF0256-17-2 500 g
DF0256-07-4 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DF0256-08-3 Supplier BD Supplier No. 225630
Add to Cart
Add to Cart
This item is not returnable. View return policy

For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles

  • Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L

Certifications

AOAC, BAM, BS12, COMPF, EP, USDA, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, BS12, COMPF, EP, USDA, USP
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Fluid Thioglycollate Medium
Product Type Dehydrated Culture Media
Formulation Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
For Use With (Application) Used for sterility testing
Packaging Type Pail
Quantity 10,000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.