Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Giolitti - Cantoni Broth Base

Catalog No. DF1809172 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF1809-17-2 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF1809-17-2 Supplier BD Supplier No. 218091
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used with Potassium Tellurite solution 3.5% selective enrichment for S. aureus isolation from food

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF, ISO
Content And Storage Refrigerate
Form Dehydrated
Media Type Giolitti-Cantoni Broth Base
Product Type Microbiology Media
Formulation Tryptone 10g/L, Beef extract 5g/L, Yeast extract 5g/L, D-Mannitol 20g/L, Sodium Chloride 5g/L, Lithium Chloride 5g/L, Glycine 1.2g/L, Sodium Pyruvate 3g/L
For Use With (Application) Used with Potassium Tellurite solution 3.5% selective enrichment for S. aureus isolation from food
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.