Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: GN Broth, Hajna

Catalog No. DF0486174 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0486-17-4 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0486-17-4 Supplier BD Supplier No. 248610
Add to Cart
Add to Cart
This item is not returnable. View return policy

For selective cultivation of gram-negative organisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BS12, CCAM, CMPH2, COMPF, MCM9
Content And Storage <30°C
Form Dehydrated
Media Type GN Broth, Hajna
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 12g/L, Proteose Peptone No. 3 8g/L, Dextrose 1g D-Mannitol 2g/L, Sodium Citrate 5g/L, Sodium Desoxycholate 0.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L
For Use With (Application) Selective cultivation of gram negative organisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.