Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: HC Agar Base

Catalog No. DF0685173 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Content And Storage:
Refrigerate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Content And Storage
DF0685-17-3 Refrigerate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0685-17-3 Supplier BD Supplier No. 268510
Add to Cart
Add to Cart
This item is not returnable. View return policy

For enumerating molds in cosmetics, Use with Tween™ 80

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Form Dehydrated
Media Type HC Agar Base
Product Type Agar
Formulation Pancreatic digest of Casein 2.5g/L, Proteose Peptone 2.5g/L, Yeast extract 5g/L, Dextrose 20g/L, Disodium Phosphate 3.5g/L, Monopotassium Phosphate 3.4g/L, Ammonium Chloride 1.4g/L, Magnesium Sulfate 0.06g/L, Chloramphenicol 0.1g/L, Sodium Carbonate 1g/L,
For Use With (Application) For enumerating molds in cosmetics Use with Tween 80
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.