Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Heart Infusion Agar

Catalog No. DF0044071 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
2 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0044-07-1 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0044-07-1 Supplier BD Supplier No. 244100
Add to Cart
Add to Cart
This item is not returnable. View return policy

General-purpose medium used in cultivation of wide range of microorganisms from variety of clinical and nonclinical specimens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Blood Agar Base
Product Type Agar
Formulation Beef heart infusion from 500g 10g/L, Tryptose 10g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Used with or without Blood for the isolation and cultivation of a wide variety of microorganisms
Packaging Type Pail
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.