Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: m Enterococcus Agar

Catalog No. DF0746170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0746-17-0 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0746-17-0 Supplier BD Supplier No. 274620
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For isolating and enumerating enterococci in water and other materials by membrane filtration or pour plate technique

  • Formulation: Tryptose 20g/L, Yeast extract 5g/L, Dextrose 2g/L, Dipotassium Phosphate 4g/L, Sodium Azide 0.4g/L, Agar 10g/L, 2,3,5-Triphenyl Tetrazolium Chloride 0.1g/L

Certifications

CE, ISO, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, ISO, SMWW
Content And Storage <30°C
Form Dehydrated
Media Type m Enterococcus Agar
Product Type Agar
Formulation Tryptose 20g/L, Yeast extract 5g/L, Dextrose 2g/L, Dipotassium Phosphate 4g/L, Sodium Azide 0.4g/L, Agar 10g/L, 2,3,5-Triphenyl Tetrazolium Chloride 0.1g/L
For Use With (Application) For isolating and enumerating enterococci in Water and other materials by membrane filtration or pour plate technique
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.