Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Nutrient Agar with MUG

Catalog No. DF0023174 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0023-17-4 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0023-17-4 Supplier BD Supplier No. 223200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fluorogenic assay for detection and enumeration of E. coli in water

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, SMD
Content And Storage <30°C
Form Dehydrated
Media Type Nutrient Agar
Product Type Nutrient Agar
Formulation Beef extract 3g/L, Peptone 5g/L, Agar 15g/L, 4-Methylumbelliferyl-beta-D-glucuronide 0.1g/L
For Use With (Application) Fluorogenic assay for the detection and enumeration of Escherichia coli in Water
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.