Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Nutrient Broth

Catalog No. DF0003070 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
2 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0003-07-0 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0003-07-0 Supplier BD Supplier No. 231000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cultivation of many species of nonfastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, SMD
Content And Storage <30°C
Form Dehydrated
Media Type Nutrient Broth
Product Type Microbiology Media
Formulation Beef extract 3g/L, Peptone 5g/L
For Use With (Application) General purpose medium for the cultivation of microorganisms with non-exacting nutritional requirements
Packaging Type Pail
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.