Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)

Catalog No. DF0369087 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0369-08-7 10 kg Pail
DF0369-17-6 500 g Bottle
DF0369-07-8 2 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DF0369-08-7 Supplier BD Supplier No. 236930
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

Applications:

  • Used with or without blood for cultivation and isolation of variety of organisms
  • Used for sterility testing

Certifications

AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Tryptic Soy Agar
Product Type Tryptic Soy Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Isolation and cultivation of non-fastidious and fastidious microorganisms
Packaging Type Pail
Quantity 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.