Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Tryptose Agar

Catalog No. DF0064174 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0064-17-4 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0064-17-4 Supplier BD Supplier No. 264300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms

Cultivation of Brucella and a variety of pathogenic microorganisms

Certifications

BAM, CCAM, COMPF

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance BAM, CCAM, COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Tryptose Agar
Product Type Agar
Formulation Tryptose 20g/L, Dextrose 1g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Cultivation of a wide variety of fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.