Learn More
BD Difco™ Disposable Inoculating Needles and Loops
Catalog No. 2203120 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Volume:
1 μL
10 μL
Product Type:
Inoculating Loop
Inoculating Needle
Color:
Light Blue
Light Green
Violet
Quantity:
1000/Pk.
250/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Product Type Color Quantity
22-031-20 1 μL Inoculating Loop Light Green 250/Pk.
22-031-24 1 μL Inoculating Needle Violet 1000/Pk.
22-031-21 1 μL Inoculating Loop Light Green 1000/Pk.
22-031-22 10 μL Inoculating Loop Light Blue 250/Pk.
22-031-23 10 μL Inoculating Loop Light Blue 1000/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-031-20 Supplier BD Supplier No. 220214
This item is not returnable. View return policy
Includes
Evans Blue Dye Certificate of Calibration with every box of 1,000 loops, confirming their accuracy and consistency
Sterile, disposable inoculating needles do not require flaming
  • Made from a unique plastic resin for controlled flexibility
  • Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
  • Free of lubricants, oils and electrostatic charges
  • Color coded
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Volume 1 μL
Material Plastic Resin
Sterility Sterile
Color Light Green
Product Type Inoculating Loop
Includes Evans blue dye certificate of calibration
Quantity 250/Pk.