Description
Includes
Evans Blue Dye Certificate of Calibration with every box of 1,000 loops, confirming their accuracy and consistency
- Made from a unique plastic resin for controlled flexibility
- Resist cutting the agar surface, yet provide sufficient firmness to pick stubborn colonies
- Free of lubricants, oils and electrostatic charges
- Color coded
Specifications
Specifications
| Disposable | Single-use |
| Volume | 10 μL |
| Sterility | Sterile |
| Material | Plastic Resin |
| Color | Light Blue |
| Product Type | Inoculating Loop |
| Quantity | 250/Pk. |