Learn More
BD Difco™ Heart Infusion Agar
Catalog No. DF0044179 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
DF0044-17-9 500 g
1 options
This item is not returnable. View return policy
Catalog No. DF0044-17-9 Supplier BD Supplier No. 244400
Add to Cart
Add to Cart
This item is not returnable. View return policy
General-purpose medium used for a wide range of microorganisms
In plates, Heart Infusion Agar can be used for primary isolation of organisms from mixed cultures. In agar slant form, it is used primarily for organism cultivation and maintenance rather than for isolation from mixed cultures.
TRUSTED_SUSTAINABILITY
Specifications
Color Beige
Form Powder
Product Type Agar
Quantity 500 g