Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ M-Green Yeast and Mold Broth

Catalog No. B11287 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11287 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B11287 Supplier BD Supplier No. 211287
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for the detection of fungi in the routine analysis of beverages

M-Green Yeast and Mold Broth is a modification of the liquid medium, M-Yeast and Mold Broth, which was developed to improve the efficiency of detection and enumeration of fungi in sugar and other materials by the membrane filter method. The revised formula contains the indicator dye, bromcresol green.
TRUSTED_SUSTAINABILITY

Specifications

Broth Type M-Green Yeast and Mold Broth
Detects Fungi
Form Solid
For Use With (Application) Detection of fungi in the routine analysis of beverage
Incubation Conditions 30° to 35°C for 2 days
Purification Method Membrane filter method
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.