Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Niacin Assay Medium

Catalog No. DF0322154 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0322-15-4 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0322-15-4 Supplier BD Supplier No. 232210
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for determining niacin concentration by the microbiological assay technique.

Niacin Assay Medium is a dehydrated medium free from nicotinic acid and its analogs but containing all other nutrients and vitamins essential for the cultivation of L. plantarum ATCC™ 8014. The addition of nicotinic acid or its analogs in specified increasing concentrations gives a growth response that can be measured turbidimetrically or titrimetrically.

TRUSTED_SUSTAINABILITY

Specifications

Color Whitish
Form Powder
Product Type Niacin Assay Medium
For Use With (Application) Determination of Niacin concentration by the microbiological assay technique
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.