Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Nitrate Broth

Catalog No. DF0268178 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0268-17-8 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0268-17-8 Supplier BD Supplier No. 226810
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Aids in the identification of aerobic and facultative anaerobic, gram-negative microorganisms via the nitrate reduction test. BD™ Difco™ Nitrate Broth is a basal medium containing potassium nitrate. Microorganisms are inoculated into the medium and then incubated.

Microorganisms may be differentiated according to their metabolism of certain substrates. The production of gas from nitrate is an important differential test for glucose-nonfermenting gram-negative bacilli. The end product of reduction depends upon the bacterial species.

  • Nitrate Broth is a basal medium containing potassium nitrate.
  • The microorganism under evaluation is inoculated into the medium and after incubation, nitrate reduction may be determined.
  • Identifies: Acinetobacter calcoaceticus 19606, Enterobacter aerogenes 13048, Escherichia coli 25922, Pseudomonas aeruginosa 27853
TRUSTED_SUSTAINABILITY

Specifications

Broth Type Nitrate Broth
Certifications/Compliance AOAC, BAM, COMPF, ISO, SMD, USDA
Color Light to medium tan
Form Dehydrated (free-flowing, homogenous)
Product Type Microbiology Media
Formulation Beef Extract 3.0g/L; Peptone 4.0g/L; Proteose Peptone (No. 3) 1.0g/L; Potassium Nitrate 1.0g/L
For Use With (Application) For identification of aerobic and facultative anaerobic gram negative microorganisms by means of the nitrate reduction test
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.