Learn More
BD Difco™ Nutrient Agar
GSA_VA
Catalog No. DF0001170 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
DF0001-17-0 500 g
1 options
This item is not returnable. View return policy
Catalog No. DF0001-17-0 Supplier BD Supplier No. 213000
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces and other materials.
  • Beef extract provides carbohydrates, vitamins, nitrogen and salts
  • Peptones provide a source of nitrogen
  • Agar is the solidifying agent
  • Consists of peptone, beef extract and agar
  • Relatively simple formulation provides the nutrients necessary for the replication of a large number of microorganisms that are not excessively fastidious
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Form Dehydrated
Quantity 500 g