Learn More
BD Difco™ Poly Antisera
Catalog No. B228371 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Product Type:
Shigella Poly Group A
Shigella Poly Group A1
Shigella Poly Group B
Shigella Poly Group D
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B228371 Shigella Poly Group D
B227761 Shigella Poly Group A1
B228351 Shigella Poly Group B
B228341 Shigella Poly Group A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. B228371 Supplier BD Supplier No. 228371
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
For agglutination test
Salmonella poly antiserum
  • Febrile control antisera
  • Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
  • Used along with febrile antigens as positive or negative controls
Shigella antiserum poly group
  • For identification of morphologically and biochemically identified Shigella species
  • Can be used only in slide agglutination test
  • Lyophilized, polyclonal rabbit antisera
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance For in vitro diagnostic use
Product Type Shigella Poly Group D
Quantity 3 mL
For in vitro diagnostic use