Learn More
MODULUS DATA SYSTEMS, INC. DIFFCOUNT™ Barcode Scanner
Catalog No. 21403168
Change view
Click to view available options
Type:
Barcode Scanner
Catalog No. Type
21-403-168 Barcode Scanner
1 options
Catalog No. 21-403-168 Supplier MODULUS DATA SYSTEMS, INC. Supplier No. 14500
  • $180.00
    $155.65 / Each of 1
    Save $24.35
    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Reduce manual data entry and input errors, and quickly scan sample identifiers with this barcode scanner.
  • Choose either handheld trigger or auto-scan modes for fast and efficient scanning of sample identifiers.
  • Supports standard 1D and 2D barcode types for versatile laboratory applications.
  • Resolution: 4 mil, Sensor: 640 x 480 CMOS for accurate and reliable barcode detection.
  • Scan speed: 500 scans/sec for quick and consistent performance.
  • Always on operation with no batteries or recharging necessary for uninterrupted workflow.
  • Includes stand and USB cable for convenient setup and connectivity.
Specifications
Type Barcode Scanner
Connection Type USB
Display Type None Included
Power Supply USB
Resolution 4 mil