Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Diffraction Grating, 300 Lines Per mm
SDPSCI_ED

Catalog No. S139470 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
9 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139470 9 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139470 Supplier United Scientific™ Supplier No. DFG300
Add to Cart
Add to Cart
This item is not returnable. View return policy

300 Lines Per mm Diffraction Grating for use with the Intermediate Spectrometer with 30 mm optics.

  • 300 Lines Per mm
  • For the Intermediate Spectrometer with 30 mm optics (SPECT03)

Specifications

Grade 9 to 15
Format Kit
For Use With (Equipment) Intermediate Spectrometer with 30 mm Optics
Product Type Diffration Grating
Languages English
Subject Chemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.