Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Diffusion and Cell Size
GREENER_CHOICESCI_ED

Catalog No. S98650 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S98650 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98650 Supplier Innovating Science™ Supplier No. IS3003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Materials for 15 groups of students, teacher's guide, student study guide copymasters

Why are cells microscopic?

Demonstrates the need for cells to effectively move materials in and remove waste.

Students will:

  • Create simulated cells (agar blocks) of different sizes
  • Examine how effectively a substance is able to diffuse into the cell in a set period of time
  • A special indicator in the cells will allow students to visualize the degree of diffusion
  • The results will clearly display the fact that a smaller volume creates a more favorable condition for the exchange of material across cell membrane

 

Compliance

Conforms to 49 CFR 173.4

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Helps students to visualize the degree of diffusion
Includes Teacher's manual and student study guide copymasters, 75g agar bottle, 25ml bromothymol blue concentrate bottle, three bottles of 25ml 2.0m hydrochloric acid, 16 agar block casting trays, 15 plastic cups, 15 plastic knives, 15 plastic stirrers
Quantity 1
Dimensions (L x W x H) 11.125 x 8.75 x 4 in.
Class Size 15 Lab Groups
Product Type Diffusion and Cell Size Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.