Learn More
Diagnocine LLC DIFFUSIVE THERMOPROBE
SDP
Supplier:  Diagnocine LLC FNKFDV0002
Encompass
Size: 100ugStorgae: RTUltrasensitive Fluorescent Thermometer for cellular temperature monitoring.The "Diffusive Thermoprobe" Useful Tools to monitor intracellular temparature
Catalog No. NC1727041