Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diflufenican 98.0+%, TCI America™
SDP

Catalog No. D410825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D410825G 25 g
D41085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D410825G Supplier TCI America Supplier No. D410825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 83164-33-4
Molecular Formula C19H11F5N2O2
Molecular Weight (g/mol) 394.301
MDL Number MFCD01311804
InChI Key WYEHFWKAOXOVJD-UHFFFAOYSA-N
Synonym diflufenican, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy nicotinamide, diflufenicanil, diflufenican bsi:iso, unii-3t8d1vdo6k, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy pyridine-3-carboxamide, 3t8d1vdo6k, 2',4'-difluoro-2-alpha,alpha,alpha-trifluoro-m-tolyloxy nicotinanilide, n-2,4-difluorophenyl-2-3-trifluoromethyl phenoxy-3-pyridinecarboxamide
PubChem CID 91735
ChEBI CHEBI:81824
IUPAC Name N-(2,4-difluorophenyl)-2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide
SMILES C1=CC(=CC(=C1)OC2=C(C=CC=N2)C(=O)NC3=C(C=C(C=C3)F)F)C(F)(F)F

Specifications

Melting Point 161°C
Quantity 25 g
Formula Weight 394.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Diflufenican
RTECSNumber US4589800
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.