Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Difluoromethoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. D491425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D491425G 25 g
D49145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D491425G Supplier TCI America Supplier No. D491425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 458-92-4
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD00068184
InChI Key LMVBQQAXGZVBFH-UHFFFAOYSA-N
Synonym alpha,alpha-Difluoroanisole
PubChem CID 68015
IUPAC Name (difluoromethoxy)benzene
SMILES FC(F)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 25 g
UN Number 1993
Formula Weight 144.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Difluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.