Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Difurfuryl Sulfide 98.0+%, TCI America™
SDP

Catalog No. D342025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D342025G 25 g
D34205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D342025G Supplier TCI America Supplier No. D342025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13678-67-6
Molecular Formula C10H10O2S
Molecular Weight (g/mol) 194.248
MDL Number MFCD00010081
InChI Key UYLKDZXJEKFFHJ-UHFFFAOYSA-N
Synonym difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide
PubChem CID 61659
IUPAC Name 2-(furan-2-ylmethylsulfanylmethyl)furan
SMILES C1=COC(=C1)CSCC2=CC=CO2

Specifications

Color White-Yellow
Boiling Point 143°C
Quantity 25 g
Formula Weight 194.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Difurfuryl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.