Learn More
Sigma Aldrich Fine Chemicals Biosciences DIG Easy HybTM Granules
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11796895001
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.A The DIG System was established as a sensitive and cost-effective alternative to using radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.
Catalog No. 50-290-5768
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.