Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Life Sciences™ DIGE Gels LF24

Catalog No. 09928200 Shop All Cytiva Products
Change view
Click to view available options
Gel Type:
Polyacrylamide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Type
09-928-200 Polyacrylamide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-928-200 Supplier Cytiva Life Sciences™ Supplier No. 29706670
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Large format precast 12.5% polyacrylamide gels with low fluorescent glass plates intended for 2D DIGE and 2D DIBE™.

DIGE Gels LF24 are large format precast 12.5% polyacrylamide gels with low fluorescent glass plates intended for 2D DIGE/DIBE™ with Amersham™ DIGE Unit LF24 electrophoresis.

  • Ready to use precast gel
  • Quick set up with optimized DIGE Buffer Kit
  • Optimized 24 cm IPG strips for large format protein separation
  • Low fluorescent glass plates enable direct fluorescent image scan
  • Suitable for large format HCP coverage assay for regulatory requirement of HCP ELISA.
  • DIGE Gels LF24 are large format 12.5% polyacrylamide precast gels with low fluorescent glass plates optimized for Amersham™ DIGE Unit LF24 electrophoresis.
  • The glass plates do not need to be removed when scanning the gel with a fluorescent image scanner. Open the glass cassette to perform transfer of the proteins to a membrane.
  • This gel is intended for two-dimensional fluorescence difference gel electrophoresis (2-D DIGE) and large format HCP coverage assay (2D DIBE™) for regulatory requirements of HCP ELISA.
  • Ready to use DIGE buffer kits allow for a quick electrophoresis set up.
  • The package contains 3 precast gels.
TRUSTED_SUSTAINABILITY

Specifications

Buffer Special Buffer Based on Piperidinopropionamide (PPA)
Gel Percentage 12.5%
Gel Type Polyacrylamide
Separation Range 12 to 120 kDa
Storage Requirements 4°C to 8°C
Product Type Polyacrylamide Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.