Learn More
Vernier™ Digital BTD Cable
SDPSCI_ED
Catalog No. S16116ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Interfaces
Catalog No. For Use With (Equipment)
S16116ND Interfaces
1 options
Catalog No. S16116ND Supplier Vernier™ Supplier No. CBBTD
Add to Cart
Add to Cart
Includes
Digital BTD Cable
This cable has bare wire on one end and a male BTD connector on the other end. It can be used to replace the cable on a BTD digital sensor by splicing it in with an existing cable. The cable is approximately 3 feet long.
  • This cable can be used to connect your homemade digital sensors to Vernier™ interfaces.
  • There is a BTD plug on on end and six bare wires on the other end.
Warranty and Services
1 year
Specifications
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Interfaces
Length (Metric) 1 m
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov