Learn More
Research Products International Corp Digital Dry Bath Incubators, Double Chamber with Dual Block, 6 1/4 x 14 x 5 Inches
SDP
Supplier:  Research Products International Corp 246321
Encompass
State of the art digital dry bath incubators are featured in single and double block styles. Digital temperature control and a removable hinged cover provide the highest level of temperature accuracy and uniformity. Incubators are supplied with either one or two Combination Flip Blocks depending of model. These unique blocks accommodate 14 x 0.5ml or 24 x 2.0ml tubes on one side and six PCR tube strips or 48 x 0.2ml tubes on the flip side. Chambers will also accept other standard size blocks with dimensions of 7 1/2 x 6 x 2 inches High.
Catalog No. 50-136-7698