Learn More
Vernier™ Digital Extension Cable
SDPSCI_ED
Catalog No. S16148ND Shop All Vernier Software Products
Change view
Click to view available options
Length (Metric):
2 m
Catalog No. Length (Metric)
S16148ND 2 m
1 options
Catalog No. S16148ND Supplier Vernier™ Supplier No. EXTBTD
Add to Cart
Add to Cart
Includes
Digital Extension Cable
Most of our sensors have cables that are 1.5 m long. There are times, however, that you may wish to have a longer cable. The Digital Sensor Extension Cable can be used with any Vernier sensor that connects to a digital channel of LabPro, CBL 2, or CBL, such as Motion Detectors and Photogates. Each cable is 2 meters in length.
  • For use with sensors that connect to a digital channel of a Vernier™ interface
  • The port will be labeled DIG1 or DIG2
  • Cable is 2 meters in length
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Educational use only
For Use With (Equipment) Digital Sensors
Length (Metric) 2 m