Learn More
Chemglass Life Sciences Digital Hotplate Stirrer Package (1/Ea): Hotplate Stirrer W/ Pt1000 Temperature Sensor, Boss Head, S.S. Vertical Support Rod, S.S. Holding Rod
SDP
Supplier:  Chemglass Life Sciences CG199750
Encompass_Preferred
Digital Hotplate Stirrer Package to Include 1 Each: CG-1997-10; Hotplate Stirrer with PT1000 Temperature Sensor, CG-1995-V-25; Boss Head, CG-1995-06; S.S. Vertical Support Rod, CG-1995-22; S.S. Holding Rod.Heavy duty round top MR Hei-Tec DIGITAL hot plate stirrer system with PT 1000 sensor. The unit features a 135mm diameter KeraDisk top that is scratch proof and chemically resistant. Hot plate includes all the leading safety control features and superior ease of use. PT 1000 temperature sensor for precise temperature control, overshoot protection and reproducible results. Digital speed control accurate to +/- 2% from 100 to 1400rpm, 115V 60Hz input. The back corner has a M10 female thread to accept a vertical support rod.
Catalog No. 50-131-1193