Learn More
United Scientific™ Digital Lux Meter
SDPSCI_ED
Catalog No. S27830 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Style:
Digital
Catalog No. Style
S27830 Digital
1 options
Catalog No. S27830 Supplier United Scientific™ Supplier No. LXM001
Add to Cart
Add to Cart
The Digital Lux Meter is a practical and economical solution for light measurements in a variety of learning situations.
  • Economical, compact solution for light measurement.
  • Readings range from 0 to 200,000 lx (0 to 18,500 fc).
  • Auto power-off after 5 minutes of inactivity to conserve battery life.
  • Threaded socket for tripod mounting.
Specifications
Material Plastic
For Use With (Application) Light Measurements
Product Type Lux Meter
Shape Rectangular
Style Digital