Description
- Measurements are accurate to 10 microns in all three directions
- Movements are monitored by a large, easy-to-read LED display module
- Resolution as precise as 10 microns
- Digital stereotaxic arm allows for:
- Memory of target coordinates
- Zeroing function at reference point
- 10-micron digital scale
- Upgrading an existing stereotaxic to a digital model will reduce the risk of human error and increase accuracy (10-rather than 100-micron resolution)
- Large LED display is also useful for surgeries performed in low light, or constrained environments, where reading the vernier scale is difficult
Options:
- Add-on Motorized Manipulator Arm and Axis StereoDrive Software
Keep existing arm and have a new one shipped - Conversion to Motorized Manipulator Arm and Axis StereoDrive Software
Send existing arm for conversion to motorized
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Size | 3 axes |
| Model | Add-on, Right |