Learn More
Stoelting™ Digital Manipulator Arm
SDP
Catalog No. 10000234 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Add-on, Left
Add-on, Right
Conversion, Left
Conversion, Right
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-234 Conversion, Left
10-000-232 Add-on, Left
10-000-233 Add-on, Right
10-000-235 Conversion, Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-234 Supplier Stoelting™ Supplier No. 51914
Add to Cart
Add to Cart
Request a Quote
Digital Stereotaxic Manipulator Arms have sealed electronic sensors, highly immune to outside interference and environmental conditions on each axis.
  • Measurements are accurate to 10 microns in all three directions
  • Movements are monitored by a large, easy-to-read LED display module
  • Resolution as precise as 10 microns
  • Digital stereotaxic arm allows for:
    • Memory of target coordinates
    • Zeroing function at reference point
    • 10-micron digital scale
  • Upgrading an existing stereotaxic to a digital model will reduce the risk of human error and increase accuracy (10-rather than 100-micron resolution)
  • Large LED display is also useful for surgeries performed in low light, or constrained environments, where reading the vernier scale is difficult
Options:
  • Add-on Motorized Manipulator Arm and Axis StereoDrive Software
    Keep existing arm and have a new one shipped
  • Conversion to Motorized Manipulator Arm and Axis StereoDrive Software
    Send existing arm for conversion to motorized
Specifications
For Use With (Application) Neuroscience
Size 3 axes
Model Conversion, Left