Learn More
World Precision Instrument Digital Microscope Camera-USBCAM152
SDP
Supplier:  World Precision Instrument USBCAM152
Encompass_Preferred
All three cameras are cased models with a rear mini-USB connector, hardware and software triggering image capture, digital zoom and a feature-rich, user based menu setup and control. Sentech USB cameras include a SDK, DirectX, Twain and Linux driver, as well as the Sentech Viewing Software.
Catalog No. 50-253-8410
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.