Learn More
Research Products International Corp Digital Orbital Shaker
SDP
Supplier:  Research Products International Corp 249255
Encompass_Preferred
Digital Orbital Shaker accommodates a vast array of containers and vessels that most typical laboratories require to shake. Features Hi-Lo sides to create tube or vessel holding patterns with elasticated cords. Platform holds assay plates, dishes, Erlenmeyer flasks, reagent bottles, irregular shaped vessels and much more. Unit instantly adjusts to weight load changes and unbalanced loads with no-stop electronic motor feedback allowing for truly reproducible results. Durable ABS construction is suitable for use in incubators (excluding C02) and cold rooms.
Catalog No. 50-238-8417
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.